Learn More
Aqua Solutions Dissolved Oxygen 3 Reagent Powder Pillows, pk/100
Supplier: Aqua Solutions HA98799
For determination of dissolved oxygen by the Winkler titration method. Sample Size: 60 mL. Pack of 100 powder pillows.