Learn More
Metrohm USA DISPOSABLE VIALS,2MM DIA,200/P
Supplier: Metrohm USA 67402070
Disposable vials, 2 mm diameter, transmission.200 lockable disposable glass vials (borosilicate) with a diameter of 2 mm for analyses of liquid samples in transmission mode. Suitable for the following Analyzers: NIRS XDS RapidLiquid AnalyzerNIRS XDS VialHeater + NIRS XDS Transmission OptiProbe Analyze
This item is not returnable.
View return policy