Learn More
TWD Scientific LLC 16x150mm Polyproylene Test Tubes. Pack of 250
SDP
Supplier:  TWD Scientific LLC DPTT16X150
Encompass_Preferred
Disposable 16x150mm polypropylene test tube. Test tubes have a maximum capacity of 15mL. 250/Pack
Catalog No. NC0444427