Learn More
Cole-Parmer Excel Scientific SPR-L-S01 disposable, 500/cs;1/pak
Supplier:  Cole-Parmer 1420110
Encompass
Sterile, disposable spreaders eliminate flaming for safe, convenient sample plating. Slightly upturned end won’t gouge agar.
Catalog No. NC9531138
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.