Learn More
Electron Microscopy Sciences Dispensing Slide
Supplier: Electron Microscopy Sciences 6172708
These thin transparent polyvinyl slides serve as a surface to precisely orient the tissue into the desired position and as a means to accurately transfer tissue to the embedding well floor. Tissue is applied face down to the end of the dispensing slide where it can be visualized from below and manipulated into position. The face that is visible will be laid onto the well floor and will ultimately be the embedded surface to be sectioned.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.