Learn More
Andwin Scientific DISP CELL SPREADER 500/CS
SDP
Supplier:  Andwin Scientific GT86653
Encompass_Preferred
The polypropylene cell spreaders are efficient tools for speading bacterial cultures in an even manner. Its unique L-shape design permits application on the entire Petri dish or plate surface. This product is presterilized, which removes the necessity of flaming or autoclaving. It is sold in a case of 500.
Catalog No. NC9626565
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.