Learn More
Roche Diagnostics DISCOVERY ChromoMap DAB Kit (RUO)
Supplier: Roche Diagnostics 05266645001
Biotin-free DAB detection kit for the chromogenic antigen localization in frozen sections or formaldehyde-fixed, paraffin-embedded tissue sections. For use with the VENTANA DISCOVERY series instruments.
This item is not returnable.
View return policy