Learn More
YB's Direct Inc. The Bio-Wipe Bag™ Containment System
SCI_ED
For simple, fast and complete cleanup and containment of infectious or hazardous spills
Supplier:  YB's Direct Inc. RR100
Encompass
Catalog No. 23-562-900
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Unique body fluid and chemical wipe and containment systems are super absorbent and able to handle up to 6 oz. (177mL) of liquid each.
  • Easy “inside out” cleanup method with two-thumbed mitt on interior of bag allows for right- or left-hand use
  • Opaque polyethylene plastic bag reduces the risk of cross-contamination in potentially infectious spills when used with protective gloves
  • Permanently sealed adhesive closure provides for safe, temporary containment and subsequent disposal in approved OSHA and/or ASHP containers
  • Packaged on a roll: 25 bags per roll and 4 rolls per case, for a total of 100 bags per case
  • Dispenser (23-562-907) sold separately