Learn More
Quantimetrix DipperPOCT UA DIPSTICK CONTROL
Supplier: Quantimetrix 140002
• Three months of room temperature stabilityand up to 3 years at 2ºC–8ºC. • Full dipstick immersion. • Zero wasted QC product