Learn More
Quantimetrix Dip&Spin, Urinalysis Dipstick/Microscopics Control, Level 1&2
Supplier: Quantimetrix 147001
New & Improved COD Crystals!•Applicable to manual & automated microscopy methods •Two controls in one•Liquid, made with human urine•18-month shelf life & open vial stability from date of manufacture when stored at 2ºC–8ºCOur most complete urinalysis control & coming soon, rod-shaped E. coli, the most commonly found bacteria in affected patient samples and beautifully engineered Calcium Oxalate Dihydrate (COD) crystals that are easily recognized by both standard and automated microscopy methodologies. Formulated with real human RBCs and WBCs, Dip&Spin makes for an excellent urinalysis dipstick and sediment combination quality control for the clinical lab.Compatible with most urine strip brands, manual microscopy methods, and automated analyzers such as: iQ200, UriSed, COBIO, Mindray EH Series. Now with values for Roche Cobas 6500.Thoughtfully redesigned packaging now includes barcodes on the labels for lot numbers and expiration dates and we’ve made it easier to identify each level.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.