Learn More
Thermo Scientific Dionex™ Combined Seven Anion Standard I
Supplier:  Thermo Scientific 056933
Encompass
Ion Standards are used for a variety of routine anion, cation, and standard determinations. Increase your productivity and reduce standard preparation errors with Thermo Scientific™ Dionex™ Ion Standards for ion chromatography. Available for routine anion and cation determinations, these single and combined ion standards are made with high purity, reagent-grade chemicals and water. Each standard is NIST traceable and includes a Certificate of Analysis.
Catalog No. NC1757919