Learn More
Dino Lite USB Handheld Digital Microscope AF4515ZTL - 1.3MP, 10x-140x Optical Magnification, Polarized Light, AMR, FLC, Long Working Distance
SDP
Supplier:  Dino Lite AF4515ZTL
Encompass_Preferred
The Dino-Lite Edge 1.3MP AF4515ZTL is an all-in-one device designed to meet most professional applications within 10x - 140x magnifications at longer working distances. The AF4515ZTL integrates premium Edge Series features AMR and FLC. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger. Polarization can be switched on/off or can be adjusted to offer full or half polarization for controlling glare. AMR displays the current magnification on screen with the included software. The new modular wireless-ready design allows for USB connection or a quick swap to wireless with a WF-20 adapter (sold separately) making it a completely cordless and portable microscope.
Catalog No. NC1740946