Learn More
Dino Lite USB Digital Microscope AM4113TL-M40 - 1.3MP, 5x - 40x Optical Magnification, Measurement, Long Working Distance
Supplier: Dino Lite AM4113TLM40
AM4113TL-M40 Dino-Lite Pro handheld digital microscope features 5x - 40x magnification range with a wider field of view and long working distance. Our Long Working Distance (LWD) feature is a lens system designed to provide magnification at a longer distance compared to typical microscopes. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.