Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, Honeywell Burdick & Jackson™

Catalog Number 60090233
Click to view available options
Quantity:
56 L
Packaging:
Returnable Steel Drum
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CHEBI:28262
methanesulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CHEBI:28262
methanesulfinylmethane
CS(C)=O

B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%

TRUSTED_SUSTAINABILITY
Quantity 56 L
Packaging Returnable Steel Drum
Grade Multipurpose solvents
Recommended Storage Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit