Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethyl Sulfoxide, 98%, Spectrum™ Chemical
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Dimethyl Sulfoxide, 98 Percent is a colorless liquid and is also known as DMSO. It is an important polar aprotic solvent thanks to its ability to dissolve both polar and on polar compounds. One of Its main features is that it is able to penetrate human skin. In the field of biology it is used in PCR as an inhibitor in secondary structures in DNA templates and DNA primers. It can also be used as a cryoprotectant, reducing ice formation in cell media and thus preventing necrosis of the cell during the freezing process
Specifications
CAS Max % | 1 |
Percent Purity | Approx.98% |
Grade | Reagent |
Quantity | 4 L |
Packaging | Amber Glass Bottle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction