Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Acetamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog Number 6003103
Click to view available options
Quantity:
20 L
Packaging:
HDPE Nowpak™
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
CAS Max % 100.0000%
Beilstein 1737614
Formula Weight 87.12
Percent Purity >99.5%
Grade HPLC
Density 0.942 g/cm3
Boiling Point 166°C
Quantity 20 L
Flash Point 64°C
Packaging HDPE Nowpak™
Chemical Name or Material Dimethyl Acetamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit