Learn More
Perkin Elmer US LLC DigiTUBEs 50 mL with RackLock, Includes Caps, Qty. 750
Supplier: Perkin Elmer US LLC N9308008
RackLock design allows for easy, one-handed closure of leak-free screw cap. All-in-one tubes eliminate the need for five separate vessels: Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.