Learn More
Chemglass Life Sciences Temperature Monitor Only, Digital, Type "K", 120V, -50 to 1200°C
Supplier: Chemglass Life Sciences CG349820
Temperature Monitor Only, Digital, Type "K", 120V, -50 to 1200°C. Temperature monitor features a built in audible alarm, Type “T” or “K” thermocouple receptacle, and a bright LED display for easy viewing. The monitor is contained in a compact stainless steel cabinet with a side-mounted clamp for securing to a 1/2” diameter support rod. It has a separate ON/OFF switch and a circuit breaker. Dimensions: 2.6” High x 3.2” Wide x 5.5" Deep.PLEASE NOTE: SUPPORT ROD, THERMOCOUPLE AND EXTENSION CORD ARE SOLD SEPARATELY.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.