Learn More
Labconco™ Digital Guardian™ 1000 Airflow Monitor
Mounts atop the enclosure in a factory-prepared location. Labconco™ Guardian™ 1000 Digital Airflow Monitor senses and alerts the operator to low airflow conditions.
Supplier: Labconco™ 3908800/EMD
Includes: Airflow sensor, face plate with circuit board, epoxy-coated steel housing, electrical cord with power pack and plug, vinyl tubing, airflow tube adapter and mounting hardware
Description
- Glacier white powder-coated steel casing
- LCD displays actual airflow in fpm or m/sec from 0 to 999 fpm (0 to 5.0 m/s)
- Audible/visual alarm alerts user to sustained low velocity condition
- Calibration instructions displayed on LCD
- LED lights display red for alarm, yellow for caution, green for normal
- Includes temperature-compensated sensor, external alarm, RS-232 port, night setback and alarm mute functions
- Adds approximately 15.2cm (6 in.) to height of enclosure
Certifications
UL, CE
Warranty and Services
One year
Specifications
Protector™ XVS Ventilation Station and Demonstration Hood | |
4.5 Kg | |
115 V, 60 Hz |
10 lb. | |
Monitor | |
LCD |