Learn More
Stanford Research Systems Inc Melting Point Apparatus MPA160
Supplier: Stanford Research Systems Inc MPA160
DigiMelt is a modern, low cost, digital melting point apparatus designed specifically for the student lab. Students learn the visual detection capillary method of melting point determination without the frustration of trying to control the oven temperature. DigiMelt rapidly heats the oven to the programmed start temperature, then ramps at the specified rate. The student carefully observes the samples and records the melting points with a touch of a button.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.