Learn More
Digi-Sense™ Digital Multimeter with Thermal Imager and Bluetooth™ Connectivity
View, store, and share real-time data from a safe distance on a smartphone
Catalog No. 13-310-499
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Get the most out of your data with our user-friendly mobile app
  • Unlimited data storage based on smart device capacity
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Built-in thermal imager quickly identifies hot spots
  • Rugged double-molded housing provides increased durability
  • Select from four color palettes: iron, rainbow, grey, or grey inverted
  • View measurements clearly on 6000-count TFT color LCD
  • Tilt stand offers easy hands-free viewing
  • IP65 waterproof rated
  • CAT IV-600 V, CAT III-1000 V safety rating
Specifications
Specifications
DC Voltage Accuracy: ±0.9% rdg ±5 digits, DC Current Accuracy: ±0.9% rdg ±5 digits, AC Voltage Accuracy: ±0.8% rdg ±5 digits, AC Current Accuracy: ±1.2% rdg ±5 digits, Capacitance Accuracy: ±2.5% rdg ±20 digits, Frequency Accuracy: ±0.9% ±5 digits, Resistivity Accuracy: ±0.5% rdg ±5 digits, Temperature Accuracy: ±1.5% rdg ±5.4°F (±1.5% rdg ±3°C)
NIST
Digital Multimeter
Electrical
Digital Multimeter with Infrared Thermometer
Varies based on measurement mode
−40° to +600°C
3 in.
2 in.
19 oz.
Rechargeable Li-ion
ABS
TFT Color LCD
Test leads, type K temperature probe, carrying case, AC power adapter/charger, rechargeable Li-ion battery, and NIST-traceable calibration.
Type-K
−40° to +1800°F
17.5 cm
85 cm
5.5 cm
540 g