Learn More
Dichloromethane, stabilized, MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 48538
Catalog No. 11-102-0082
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Agriculture and Environmental
Specifications
Specifications
75-09-2
MFCD00000881
84.93
Analytical Standard
Ampule
2.9 (vs. air)
-97°C
5000 mg
CH2 Cl2
Methylene chloride
Neat
1223°F
24.45 psi (55°C)
1.325 g/mL (at 25°C)
39.8°C to 40°C
Organic Pollutant Standards
Safety and Handling
Safety and Handling
RTECSNumber : PA8050000