Learn More
Biorex Labs Test Kit DIAZO-CHEK Hepatic / General Chemistry Bilirubin Urine Sample 100 Tests CLIA Waived
Supplier: Biorex Labs B1009
The DIAZO-CHEK test is used to qualitatively detect bilirubin in urine or confirm the positive results obtained by a reagent strip. 100 tests/Kit which includes precut test pad squares and reagent discs. Results are obtained in 1 minute.