Learn More
Lab-Aids™ Dialysis Experiment Kit
Observe diffusion of a solute through a semi-permeable membrane, remove an inorganic salt from a colloid and test for presence of chloride ions and starch with simple color tests.
Supplier: Lab-Aids™ 23
Includes: Kit provides sufficient materials for thirty individual experiments: 12 Chemplates™ (each with a spatula); 30 dialysis membrane tubes; 30 plastic containers; two 170g bottles of sodium chloride; a 28g bottle of silver nitrate; two 170g bottles of starch suspension; a 56g bottle of iodine solution; a 28g bottle of deionized water; 30 student instructions/worksheets; instructions and background information. Bottles are drop-controlled.
Description
Chemical and biological theories come to life in this natural continuation to the study of osmosis and diffusion. In this activity, students will:
- Use a colloidal solution of starch and sodium chloride to vividly observe dialysis
- Observe diffusion of a solute through a semi-permeable membrane
- Remove an inorganic salt from a colloid
- Test for the presence of chloride ions and starch with simple color tests
Invidualized worksheets guide students through the step-by-step procedure in this self-directed lab activity.
Specifications
Observe dialysis and diffusion of a solute through a semipermeable membrane | |
12 chemplates™ 30 dialysis membrane tubes, 30 plastic containers, two 170g bottles of sodium chloride, a 28g bottle of silver nitrate, two 170g bottles of starch suspension, a 56g bottle of iodine solution, a 28g bottle of deionized wate | |
Dialysis Experiment Kit | |
30 Students |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.