Learn More
Fisherbrand™ Fabric ESD Chair - Desk Height with Medium Back, Adjustable Arms, and ESD Casters in ESD Fabric
SDPSCI_ED
Fisherbrand™ Fabric ESD Chairs with Medium Back are height for 31 in. work surfaces & feature ergonomic adjustments, electro-static components such as fabric & self-braking casters, & arms. Chairs with Medium Back are manufactured to ensure proper grounding for static sensitive applications.
Catalog No. S40236
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Medium Back
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
ESD Chair
136 kg
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Standard
No
42 lb.
Yes
No
300 lb.
Yes
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
Polished Cast Aluminum
Yes
ANSI/ESD-STM-12.1-2006
19 to 24 in.
Yes
Blue
Polyurethane foam, heavy-duty ESD fabric
Desk Height
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov