Learn More
Sterilis Solutions Deodorizing Solution
Sterilis Solutions™ Deodorizing Solution formulated to eliminate regulated medical waste odors
Supplier:  Sterilis Solutions 38232733
 View more versions of this product
Catalog No. 23-777-014
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Environmentally friendly odor neutralizing solution
  • Designed to eliminate a wide spectrum of odors associated with Regulated Medical Waste (RMW)
  • When used as directed the Hydroponic - 50% Concentrated solution is safe for use around people, pets, and plants
  • The chemistry behind the Oxidative - 100% Concentrated solution activates free Iodine (I2) which oxidizes or dismantles organic molecules upon contact in the sterilization airstream
  • Sterilis Deodorizing Solution is not a masking agent, but neutralizes odors naturally at a molecular level and is designed to work within the Sterilis Solutions Autoclaving/Grinding system
Specifications
Specifications
Liquid
Autoclavable
Sterilis Solutions System
Unscented
Oxidizing, 100% Concentrated
1 gal.
Product Suggestions
Product Suggestions