Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Denhardt' Reagent (50X Solution/Molecular Biology), Fisher BioReagents
Description
Specifications
Denhardt's Reagent | |
Colorless | |
26873-85-8, 9003-39-8, 9048-46-8 | |
0°C | |
<1% each of Ficoll™, Polyvinylpyrrolidone, and Bovine Albumin; and Water. Each 5mL of 50X Solution contains 50mg each of Ficoll™, Polyvinylpyrrolidone, and Bovine Albumin. | |
Report (at 25°C) | |
5 mL |
0.60 to 0.80 AU at 278nm | |
50X Solution | |
100°C | |
Molecular Biology | |
Amber Glass | |
5 to 7 | |
Liquid |
Safety and Handling
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction