Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Denhardt's Reagent, 50X Powder, Molecular Biology, Fisher BioReagents
Description
Amber Glass; 150mg
- Used to make a 50X concentrate solution that can be diluted for use as a blocking agent in nucleic acid hybridization techniques
- DNase, RNase, and protease free powder that is pre-mixed for your convenience to make 500mL of 50X solution
- Eliminates the need to separately weigh ingredients - simply dissolve powder in pure water
- The 50X concentrated solution can be diluted for use in hybridization techniques
- Commonly used as a blocking buffer for Southern and Northern blot membranes
Specifications
Denhardt's Reagent | |
50X Powder | |
Denhardt's Solution | |
50 mg each of bovine serum albumin (BSA), polyvinylpyrrolidone, and Ficoll™400 | |
Report | |
150 mg |
0.6 to 0.8 AU at 278nm | |
26873-85-8, 9003-39-8, 9048-46-8 | |
Molecular Biology | |
Amber Glass | |
5 to 7 | |
Solid |
Safety and Handling
Recommended Storage : 2°C to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction