Learn More
WASP Delta WASP 2040 Clay
The Delta WASP 2040 Clay is a professional, compact LDM 3D printer that is perfect for your studio or classroom.
Specifications
Product Type | 3D Printer |
---|
Alert:
Small Parts - Not recommended for children under 3 years old.
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
S4131942
|
WASP
WU9001 |
No Support Package | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
WASP presents an LDM printer perfect for your studio or classroom.- Professional, compact 3D printer
- 20 cm (7.87 in.) base and height up to 40 cm (15.75 in.)
- Open and accessible on all three sides to interact with your print.
- Its aluminum and steel structure guarantees stability and precision, obtaining quality prints with dense fluid materials.
- Extend your warranty and add a support package for training, design/slicing consultation, and more.
Standard Configuration:
- WASP LDM 3.0 Extruder
- 1.5 mm, 2 mm, and 3 mm Diameter Nozzles
- 3 L Tank
Specifications
3D Printer |