Learn More
Roboz Surgical DELICATE BONE TRIMMERS 7MM
Supplier: Roboz Surgical RS8469
Delicate Bone Trimmer 7mm Cutting Edge, 5.5 in/ 14 cm length This delicate bone trimmer is the finest available bone cutter. The tips are thinner (only 0.85mm) and the tip profile is longer and pointier than other bone cutters or even scissors