Learn More
QUAD FIVE DEFIBRINATED SHEEP BLOOD 500ML
Supplier:  QUAD FIVE 610500
Encompass
Defibrinated Sheep Blood 500ml. Blood is mechanically defibrinated in the original collection bag and pooled before final filling. Ship bags are gamma irradiated and equipped with a filter and luer lock connectors. Volumes can be customized to your batch size. Oxygen can be added to bags upon request. Washed Red Blood Cells in 0.85% Saline.
Catalog No. NC0347755