Learn More
Utak Defibrinated Plasma, 500mL
Supplier: Utak 44600D(500)
100% Real defibrinated plasma is collected from healthy adults in FDA-licensed facilities within the United States. The defibrinated plasma is screened for common drugs of abuse and alcohol. This product contains no preservatives and is suitable for all testing methods, including LCMS, LC-MS/MS, GCMS, Time of Flight (TOF), ELISA, EMIT and other immunoassays.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.