Learn More
QUAD FIVE DEFIBRATED SHEEP BLOOD 250ML
Supplier:  QUAD FIVE 610250
Encompass
DONOR STERILE DEFIBRINATED WHOLE SHEEP BLOOD, 250 ml, Hematocrit: > 35%, Osmotic Fragility: 0% hemolysis in 0.65% saline solution
Catalog No. NC9506700