Learn More
Fisherbrand™ DeepWell™ 96-well Polypropylene Microplates PROMO
Ideal for storage because of the low-binding characteristics of polypropylene.
Description
- Accommodate even larger working volumes; up to 2 mL storage volume per well
- Proteins and DNA will not adhere to polypropylene, allowing for complete sample recovery
- Natural color
- Non-sterile and sterile available
Specifications
U | |
8 x 12 | |
Polypropylene | |
96 | |
Round |
Standard | |
Natural | |
2 mL | |
Non-sterile | |
60 Microplates |