Learn More
VAI DECON-PHENE PLUS Non-Sterile, 1 Gallon SimpleMix
A high pH phenolic that is compatible with most non-porous hard surface materials and can be used on surfaces in aseptic filling and gowning rooms, in general manufacturing areas, and in laboratories.
$205.00 - $221.00
Specifications
Product Type | Phenolic Disinfectant |
---|---|
Form | Clear Liquid |
Scent | Unscented |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Dilution | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dilution | Price | Quantity & Availability | |||||
19-546-438
|
VAI
DPP051Z |
1:128 |
Case of 4 for $205.00
|
|
|||||
19-546-439
|
VAI
DPP052Z |
1:64 |
Case of 4 for $221.00
|
|
|||||
Description
- Effective sanitizer on inanimate, non-porous, non-food contact surfaces
- Sterile pharmaceutical cleanroom formula
- Phosphate free
Specifications
Phenolic Disinfectant | |
Clear Liquid | |
Unscented | |
Non-sterile |