Learn More
Rotronic Instrument Corp Data Logger, Wireless Interface, 915 mHz
Supplier:  Rotronic Instrument Corp RMSLOG915
Encompass_Preferred
The data logger is the flexible component between the probe and the database in the ROTRONIC Monitoring System. It stores 44,000 pairs of measured values from the exchangeable HygroClip probe, and transmits them to the RMS database via LAN or wireless link. It guarantees absolute data protection, even if power supply and communications should temporarily break down.
Catalog No. NC1930225