Learn More
Pendotech Pendotech Pressuremat And Cmont Data Acquisition And Trending Software
Supplier: Pendotech PMATPGUI
Data Acquisition and Trending Software for PressureMAT and CMONT with 2 USB/serial cables to connect to a PC