Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Dade Thrombin Reagent
Determination of fibrinogen in plasma and to accelerate coagulation of anticoagulated samples for immunohematology studies.
$202.00 - $612.00
Specifications
Type | Hemostasis Reagent |
---|---|
For Use With (Application) | Fibrinogen Testing |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-044-655
|
Siemens Healthineers
10445720 |
10 x 1 mL |
Pack of 10 for $202.00
|
|
|||||
23-044-656
|
Siemens Healthineers
10445721 |
10 x 5 mL |
Pack of 10 for $612.00
|
|
|||||
Description
Dade Thrombin Reagent is an effective reagent for use in the determination (Clauss method) of fibrinogen in the detection of hereditary or acquired hypo- and hyperfibrinogenemia, dysfibrinogenemia, and afibrinogenemia. The reagent offers long stability after reconstitution.Specifications
Hemostasis Reagent | |
Fibrinogen Testing |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title