Learn More
Cole-Parmer Digi-Sense ASTM S63C-03 Individually Calibrated Liquid-In-Glass Thermometer; -8/32C, Organic Liquid Fill
Supplier: Cole-Parmer 0800966
These NIST-traceable thermometers fulfill requirements for performing ASTM methods without the worry of mercury contamination. Organic fill solution is safer to clean up in the event of breakage. The calibration report states corrections to one-tenth of the smallest division, providing an accurate offset for ensuring precise measurement records.