Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Glutamine, 99.2%, MP Biomedicals™
Click to view available options
10g
Specifications
CAS | 5959-95-5 |
Color | White |
Assay Percent Range | 0.992 |
Molecular Formula | C5H10N2O3 |
Synonym | d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln |
InChI Key | ZDXPYRJPNDTMRX-GSVOUGTGSA-N |
SMILES | C(CC(=O)N)C(C(=O)O)N |
IUPAC Name | (2R)-2,5-diamino-5-oxopentanoic acid |
Molecular Weight (g/mol) | 146.146 |
PubChem CID | 145815 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction