Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Arabinose (White Powder), Fisher BioReagents

Click to view available options
Amber Glass; 100g
D-(-)-Arabinose is a component used for culture media.
CAS 28697-53-2
Melting Point 152°C
Color White
Assay Percent Range ≥99 %
Molecular Formula C5H10O5
MDL Number MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
Packaging Amber Glass
Quantity 100 g
Synonym alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
InChI Key PYMYPHUHKUWMLA-DEOSMSJNSA-N
SMILES OCC(O)[C@@H](O)[C@H](O)C=O
Specific Rotation -107 to -97° (+ or -)
IUPAC Name (4S,5R)-oxane-2,3,4,5-tetrol
Molecular Weight (g/mol) 150.13
PubChem CID 71313478
Formula Weight 150.07
Percent Purity 99% min.
Physical Form Powder or Solid
Chemical Name or Material D-(-)-Arabinose
Show More Show Less

Recommended Storage : RT

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit