Learn More
Sartorius Incucyte Cytolight Lentivirus (EF1a, Puro), Red, 0.6 mL
Supplier:  Sartorius 4482
Encompass
One vial of lentivirus reagent (0.6mL) suitable for transducing cells with non-perturbing cytoplasmic fluorescent label. Stable cell lines can be created under puromycin selection.
Catalog No. NC0844349
Due to product restrictions, please to purchase or view availability for this product.