Learn More
Agilent BioTek Cytation 5 Multi-Mode Only Reader, IVD
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For In Vitro Diagnostic Use.
$43737.75 - $92280.30
Specifications
Type | Multi-Mode Only Microplate Reader |
---|---|
Shaking | Linear, Orbital, Double Orbital |
CO2 Concentration Range | 0 to 20% |
Absorbance Range | 230 to 999 nm, 1 nm increments |
Accuracy | Monochromator: ±2 nm; Dispenser: ±1 μL or 2% |
Catalog Number | Mfr. No. | Model | Plate Type | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Plate Type | Includes | Price | Quantity & Availability | |||
BTCYT5FSI
|
Agilent BioTek
CYT5FSI |
CYT5F | Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates | Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence |
Each for $43,737.75
|
|
BTCYT5FASI
|
Agilent BioTek
CYT5FASI |
CYT5FA | Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates | Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence/alpha detection |
Each for $52,065.30
|
|
BTCYT5MSI
|
Agilent BioTek
CYT5MSI |
CYT5M | Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible | Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence |
Each for $63,152.25
|
|
BTCYT5MFSI
|
Agilent BioTek
CYT5MFSI |
CYT5MF | Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible | Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence; Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence |
Each for $83,953.80
|
|
BTCYT5MFASI
|
Agilent BioTek
CYT5MFASI |
CYT5MFA | Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible | Filter-based fluorescence intensity/fluorescence polarization/time resolved fluorescence/luminescence/alpha detection; Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence |
Each for $92,280.30
|
|
Description
Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. The microscopy module offers up to 60x magnification in fluorescence, brightfield, high contrast brightfield, color brightfield, and phase contrast to address many applications and workflows
The multi-mode detection modules include filter- and monochromator-based fluorescence detection, luminescence, and UV-Vis absorbance detection. Gen5 software provides complete control over all imaging and data capture, plus powerful image and data analysis.
Specifications
Multi-Mode Only Microplate Reader | |
0 to 20% | |
Monochromator: ±2 nm; Dispenser: ±1 μL or 2% | |
BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible | |
2 Syringe Pumps | |
Xenon Flash; 100 mW 680 nm laser (Alpha Detection) | |
4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C) | |
51.4 cm | |
44.5 cm | |
36.3 kg | |
41.6 cm | |
250 W max. |
Linear, Orbital, Double Orbital | |
230 to 999 nm, 1 nm increments | |
Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades | |
5 to 1000 μL, 1 μL increments | |
End point, kinetic, spectral scanning and well area scanning | |
Gen5 Microplate Reader and Imager Software included; Gen5 Image+ and Image Prime software available for full image analysis; Gen5 Secure for 21 CFR Part 11 compliance (option) | |
20.2 in. | |
17.5 in. | |
80 lb. | |
16.4 in. | |
CE and TUV marked. RoHS compliant. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.