Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyproheptadine hydrochloride, Tocris Bioscience™

Catalog Number 099650
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
969-33-5
C21H22ClN
323.86
MFCD00012538
ZPMVNZLARAEGHB-UHFFFAOYSA-N
cyproheptadine hydrochloride, cyproheptadine hcl, cipractin, nuran, periactin hydrochloride, anarexol, antegan, periactin syrup, peritol, component of dronactin
13770
CHEBI:59695
hydrogen 1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine chloride
[H+].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12
This item is not returnable. View return policy
969-33-5
C21H22ClN
323.86
MFCD00012538
ZPMVNZLARAEGHB-UHFFFAOYSA-N
cyproheptadine hydrochloride, cyproheptadine hcl, cipractin, nuran, periactin hydrochloride, anarexol, antegan, periactin syrup, peritol, component of dronactin
13770
CHEBI:59695
hydrogen 1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine chloride
[H+].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12

5-HT2 antagonist

Non-selective 5HT2 antagonist, migraine prophylactic. Also SETD7/9 inhibitor (IC50 = 1μM). Decreases expression and increases degradation of estrogen receptor (ER) α in breast tumor MCF7 cells. Inhibits growth of MCF7 xenografts in mice.
Quantity 50 mg
Formula Weight 323.86
Percent Purity >98%
Chemical Name or Material Cyproheptadine hydrochloride
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit