Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanone, Honeywell Riedel-de Haen™

Catalog Number 60090232
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
108-94-1
7967
CHEBI:17854
108-94-1
7967
CHEBI:17854

Puriss., ≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Percent Purity min. 99.5 %
Grade Reagent
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Water Max. 0.2 %
Recommended Storage Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit