Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Honeywell

Catalog Number 6000484
Click to view available options
Quantity:
204 L
Packaging:
Metal Drum
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
CAS Max % 100.0000%
UN Number UN1145
Beilstein 1900225
Solubility Information 0.6% in Water at 20°C
Formula Weight 84.16
Percent Purity >99%
Grade ACS Reagent
Density 0.78 g/cm3
Boiling Point 80.72°C
Quantity 204 L
Flash Point −20°C
Packaging Metal Drum
Water 0.02% max.
Assay by GC Analysis
Chemical Name or Material Cyclohexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclohexane
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit