Learn More
Epredia™ CTM6 Coverslipper
Time-saving efficiency, user safety and compact form help make the Epredia™ CTM6 Coverslipper a good choice for any laboratory.
Supplier:  Epredia™ 970010
Catalog No. 97-001-0
Add to Cart
Description
Description
  • Efficiency for Time Savings - Coverslip hundreds of slides per hour with self-cleaning dispensing needle
  • Change to different sizes and styles of coverslips quickly and easily for versatility
  • User Safety - Load and unload slide racks without opening the coverslipper cover
  • Internal vapor extraction system with permanent air-flow avoids user exposure
  • Intergrated charcoal filtration
  • Tool-free cleaning and maintenance
  • Compact Form - Designed to fit almost any lab space with a 30 inch or larger hood space
Specifications
Specifications
Automated Coverslipper
44.1 lb.
50/60 Hz
45 x 46 x 35.5 cm (17.7 x 18.1 x 14 in.)
20 kg
100/240 V