Learn More
Thermo Scientific CSG16 50X3.0 MM
Supplier:  Thermo Scientific 079931
Encompass
Determine disparate concentration ratios of adjacent eluting cations such as sodium and ammonium in diverse sample matrices using the Thermo Scientific™ Dionex™ IonPac™ CS16 Capillary, Analytical & Guard Column. Sample matrices include environmental waters; power plant waters treated with ammonium, morpholine or ethanolamine; chemical additives; chemical process solutions; scrubber solutions; plating baths; and industrial solvents. The Dionex IonPac CS16 column is designed and recommended for use in most Thermo Scientific™ Dionex™ IonPac™ CS15 column applications due to its larger linear working range.
Catalog No. NC0399437