Learn More
Thermo Scientific Analytical Column 4mm x 250mm
Supplier: Thermo Scientific 076026
Dionex™ IonPac™ CS19 IC Column 4mm x 250mm, Max. Pressure: 3000 psi (207 bar), Separate polar amines with the Thermo Scientific™ Dionex™ IonPac™ CS19 Cation-Exchange Column. This high-capacity, carboxylate-functionalized column is specifically optimized for the separation of common cations, small polar amines, and moderately hydrophobic and polyvalent cations using simple aqueous eluents and elevated temperatures. The column is ideal for analysis of power plant waters treated with ammonium, morpholine, or ethanolamine and can also be used to analyze chemical additives, chemical process solutions, scrubber solutions, personal care products, and food samples.