Learn More
Epredia™ Cryotome Cryocassettes
Use Epredia™ Cryotome™ Cryocassettes with FSE/FE and SME Cryotomes™.
Supplier: Epredia™ 77210154
Description
Product Note:- The Cryotome FSE Cryostat has specimen holder components pre-mounted in cryochamber (cryocassette clamp, XY orienting head, and Peltier). These are replaceable only through manufacturer service.
Specifications
Variety pack (one each) | |
FSE/FE/SME |